missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog No. AC192850050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192850050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC192850050 Supplier Thermo Scientific Chemicals Supplier No. 192850050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Pink to Violet
Melting Point 1495.0°C
Boiling Point 2870.0°C
Quantity 5 g
Packaging Glass bottle
Form Powder
Assay Percent Range 99.995%
Formula Weight 58.93
Percent Purity 99.995%
Grade Trace Metal Basis
Chemical Name or Material Cobalt
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Flammable solid. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed.
Precautionary Statement GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

RUO – Research Use Only