Learn More
BINDER™ Coated Heat Exchanger
Heat exchanger that is coated in plastic to prevent damage.
Supplier: BINDER™ 80122464
Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
| KB PRO 720, KB PRO 1060 | |
| Aluminum | |
| Heat Exchanger |