Learn More
BINDER™ Coated Heat Exchanger
Heat exchanger that is coated in plastic to prevent damage.
Supplier: BINDER™ 80122586
Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
| KBF PRO 1600 | |
| Aluminum | |
| Heat Exchanger |