Learn More
Coal tar, Thermo Scientific Chemicals
$185.36 - $185.36
Description
Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 8007-45-2 | |
| MFCD00146482 | |
| Tar |
Specifications
| 8007-45-2 | |
| MFCD00146482 | |
| 14,2426 | |
| Not miscible or difficult to mix with water. | |
| Coal tar |
| Petroleum-like | |
| 500 mL | |
| Tar | |
| Viscous Liquid |
Safety and Handling
GHS H Statement
H350
May cause cancer.
P201-P202-P281-P308+P313-P501c
H350
EINECSNumber : 232-361-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only