missing translation for 'onlineSavingsMsg'
Learn More

Coal tar, Thermo Scientific Chemicals

Numéro de catalogue. AA42488AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA42488AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AA42488AP Fournisseur Thermo Scientific Chemicals Code fournisseur 042488AP
Il en reste null

Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 8007-45-2
MDL Number MFCD00146482
Synonym Tar

Spécifications

Odor Petroleum-like
Quantity 500 mL
Merck Index 14,2426
Solubility Information Not miscible or difficult to mix with water.
Physical Form Viscous Liquid
Chemical Name or Material Coal tar
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 232-361-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only