missing translation for 'onlineSavingsMsg'
Learn More

Coal tar solution, contains 81.0-86.0% alcohol

CAS: 8007-45-2

$97.81 - $379.53

Chemical Identifiers

CAS 8007-45-2
MDL Number MFCD00146482
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43671AP
View Documents
Thermo Scientific Chemicals
043671AP
500 mL
Each for $97.81
Only null left
 
AA43671K4
View Documents
Thermo Scientific Chemicals
043671K4
2.5 L
Each for $379.53
Only null left
 
Description

Description

Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis). Coal tar has mild antiseptic and anti-inflammatory effects, and relieves itching. It is also absorbed into the skin cells and is thought to inhibit DNA replication in these cells. This slows down cell division and stops the cells from multiplying excessively. In turn, this reduces the thickening and scaling of the skin and clears up psoriasis plaques. It also finds its application in surface coatings and sealants. Also used as a fuel in open hearth furnaces in the steel industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8007-45-2
MFCD00146482

Specifications

8007-45-2
13°C (55°F)
MFCD00146482
UN1993
Contains 81.0 to 86.0% alcohol
Coal tar solution
Brown
Alcohol-like
500 mL
Fully miscible in water.
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H350
Highly flammable liquid and vapor.
May cause cancer.

P201-P202-P210-P233-P240-P241-P242-P243-P264b-P280i-P281-P303+P361+P353-P305+P351+P338-P308+P313-P370+P378q-P501c

H225-H319-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

For Manufacturing and Laboratory Use. Not for beverage use