missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution

Catalog No. 22040119
Change view
Click to view available options
Capacity (Metric):
2 mL
2.7 mL
Closure Color:
Blue with Black Ring
Blue with White Ring
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22040119 2.7 mL Blue with Black Ring
22040045 2 mL Blue with White Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22040119 Supplier Greiner Bio-One Supplier No. 454334

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For routine hemostasis testing

  • 13 x 75 mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull

Specifications

Product Type Sodium Citrate Blood Collection Tube
Additive Buffered Sodium Citrate, 3.2%
Material PET
Capacity (Metric) 2.7 mL
Closure Color Blue with Black Ring
Tube Size 13 x 75 mm