Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
Supplier: Fisherbrand™ 332353G04
Description
- Maintains temperature down to -67°C with CO2
Specifications
| Factory Installed | |
| -67°C | |
| Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer |