Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Supplier:  Thermo Scientific™ FLN300W
 View more versions of this product
Catalog No. FLN300W
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
Specifications
LN2 Backup, Standalone
230V/50Hz models (UK Plug): Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086
230 V 50 Hz
Boxes (300)
-80°C
230 V
50 Hz
Product Suggestions
Product Suggestions