Learn More
Cytiva Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA suspended screen
Catalog No. 5000053018
Encompass
Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA suspended screen
Catalog No. 5000053018 Supplier Cytiva Supplier No. PSM65C11P1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Centramate cassette Supor PES membrane 0 65 m removal rating 0 01 m EFA suspended screen