missing translation for 'onlineSavingsMsg'
Learn More

Clozapine

CAS: 5786-21-0 | C18H19ClN4 | 326.83 g/mol

$578.63 - $578.63

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.83
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
IUPAC Name 6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
SMILES CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC464280010
View Documents
Thermo Scientific Chemicals
464280010
1 g
Each for $578.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5786-21-0
326.83
6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
C18H19ClN4
ZUXABONWMNSFBN-UHFFFAOYSA-N
CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1

Specifications

5786-21-0
C18H19ClN4
Solubility aq. soln.: insoluble.
CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
326.83
Clozapine
182.0°C to 185.0°C
1 g
ZUXABONWMNSFBN-UHFFFAOYSA-N
6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
326.82
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Suspected of causing genetic defects.

Suspected of damaging fertility or the unborn child.<b

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 227-313-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only