missing translation for 'onlineSavingsMsg'
Learn More

Clozapine 98.0+%, TCI America™

Catalog No. c25471g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
100 mg
Catalog No. Quantity
C25471G 1 g
C2547100MG 100 mg
2 options

Catalog No. C25471G

Supplier: TCI America C25471G

Only null left

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.828
MDL Number MFCD00153785
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
Synonym clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
PubChem CID 2818
IUPAC Name 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl

Specifications

Melting Point 185°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 326.83
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Clozapine
RTECSNumber HP1750000
TSCA No