missing translation for 'onlineSavingsMsg'
Learn More

Clozapine N-oxide, 99%, Thermo Scientific Chemicals

Catalog No. AAJ67235LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67235LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67235LB0 Supplier Thermo Scientific Chemicals Supplier No. J67235LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34233-69-7
Molecular Formula C18H19ClN4O
Molecular Weight (g/mol) 342.827
MDL Number MFCD00210190
InChI Key WYRDWWAASBTJLM-UHFFFAOYSA-N
Synonym clozapine n-oxide, clozapine-n-oxide, unii-mza8bk588j, chembl1688, mza8bk588j, n-oxyclozapine, clozapine n-oxide cno, oguczbiqsyywef-uhfffaoysa-n
PubChem CID 2819
IUPAC Name 3-chloro-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES C[N+]1(CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl)[O-]

Specifications

Color Yellow
Quantity 5 mg
UN Number UN2811
Solubility Information Soluble in DMSO.
Formula Weight 342.83
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Clozapine N-oxide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber HP1760000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only