missing translation for 'onlineSavingsMsg'
Learn More

Clozapine, 97%

Catalog No. AAJ61583MB
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61583MB 25 mg
AAJ61583MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61583MB Supplier Thermo Scientific Chemicals Supplier No. J61583MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potent, selective muscarinic antagonist | CAS: 5786-21-0 | C18H19ClN4 | 326.828 g/mol

Potent, selective muscarinic antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.828
MDL Number MFCD00153785
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
Synonym clozapine, leponex, clozapin, clozaril, fazaclo, clorazil, iprox, clozapinum, asaleptin, clozapina
PubChem CID 2818
IUPAC Name 3-chloro-6-(4-methylpiperazin-1-yl)-5H-benzo[b][1,4]benzodiazepine
SMILES CN1CCN(CC1)C2=C3C=CC=CC3=NC4=C(N2)C=C(C=C4)Cl

Specifications

Melting Point 182°C to 184°C
Color Yellow
Quantity 25 mg
UN Number UN2811
Merck Index 14,2423
Formula Weight 326.83
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Clozapine
Hazard Category H301-H315-H319-H335-H341-H361
Hazard Statement GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 227-313-7
RTECSNumber HP1750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only