missing translation for 'onlineSavingsMsg'
Learn More

Cloxacillin Sodium Salt Monohydrate 98.0+%, TCI America™

Catalog No. C22445G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22445G 5 g
C22441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22445G Supplier TCI America Supplier No. C22445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
MDL Number MFCD00150735
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
Synonym cloxacillin sodium, cloxapen, bactopen, cloxacillin sodium monohydrate, sodium cloxacillin monohydrate, tegopen, staphobristol-250, orbenin sodium hydrate, cloxacap, orbenin
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 457.86
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cloxacillin Sodium Salt Monohydrate
RTECSNumber XH8750000
TSCA Yes