missing translation for 'onlineSavingsMsg'
Learn More

Clove Oil, FCC, 85%, Spectrum™ Chemical

Catalog No. p-7157277
Change view
Click to view available options
Quantity:
25 mL
125 mL
15 kg
Packaging:
Amber Glass Bottle
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601769 25 mL Amber Glass Bottle
18601770 125 mL Amber Glass Bottle
18601772 15 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601769 Supplier Spectrum Chemical Mfg Cor Supplier No. CL13025ML
Only null left

CL130, 8000-34-8

Spectrum™ Chemical Clove Oil, FCC is a flavor and fragarant ingredient. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8000-34-8
Molecular Formula C7H12ClN3O2
Molecular Weight (g/mol) 205.64
InChI Key VEEIFXWJNCAVEQ-UHFFFAOYNA-N
IUPAC Name methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CN=CN1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.527-1.535
Quantity 25 mL
Percent Purity 85%
Grade FCC