missing translation for 'onlineSavingsMsg'
Learn More

Clomipramine Impurity F, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022975
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022975 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022975 Supplier Merck Emd Millipore Supplier No. BP983

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 32943-25-2
Molecular Formula C14H12ClN
Linear Formula C14H12ClN
Quantity 10 mg
Synonym 3-Chloro-10,11-dihydro-5 H-dibenzo[b,f]azepine
Formula Weight 229.70
Grade BP