missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clomipramine Impurity D, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 3589-22-8 |
| Molecular Formula | C19H22Cl2N2 |
| Linear Formula | C19H22Cl2N2 |
| Quantity | 10 mg |
| Synonym | 3-(3,7-Dichloro-10,11-dihydro-5 H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine |
| Formula Weight | 349.30 |
| Grade | British Pharmacopoeia (BP) Reference Standard |