missing translation for 'onlineSavingsMsg'
Learn More

Clomipramine hydrochloride

Catalog No. AAJ6248506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6248506 5 g
AAJ6248503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6248506 Supplier Thermo Scientific Chemicals Supplier No. J6248506

Potent, selective 5-HT uptake blocker | CAS: 17321-77-6 | C19H24Cl2N2 | 351.315 g/mol

Potent, selective 5-HT uptake blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17321-77-6
Molecular Formula C19H24Cl2N2
Molecular Weight (g/mol) 351.315
MDL Number MFCD00069234
InChI Key WIMWMKZEIBHDTH-UHFFFAOYSA-N
Synonym clomipramine hydrochloride, anafranil, clomipramine hcl, 3-3-chloro-10,11-dihydro-5h-dibenzo b,f azepin-5-yl-n,n-dimethylpropan-1-amine hydrochloride, anaphranil, 3-chloroimipramine hydrochloride, chloroimipramine monohydrochloride, chlorimipramine hydrochloride, unii-2lxw0l6gwj
PubChem CID 68539
ChEBI CHEBI:3755
IUPAC Name 3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl.Cl

Specifications

Melting Point 189°C to 190°C
Color White
Quantity 5 g
Merck Index 14,2387
Formula Weight 351.32
Physical Form Powder
Chemical Name or Material Clomipramine hydrochloride
Hazard Category H302+H312+H332-H360
Hazard Statement GHS H Statement
H360-H302-H312-H332
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 241-344-3
RTECSNumber HN9055000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only