missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clofazimine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 219°C |
| Color | Red-Yellow |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 473.40 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | Clofazimine |
Safety and Handling
| RTECSNumber | SG1578000 |
| TSCA | No |