missing translation for 'onlineSavingsMsg'
Learn More

Clofazimine 98.0+%, TCI America™

Catalog No. C28661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28661G 1 g
C28665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28661G Supplier TCI America Supplier No. C28661G
Only null left

Chemical Identifiers

CAS 2030-63-9
Molecular Formula C27H22Cl2N4
Molecular Weight (g/mol) 473.401
MDL Number MFCD00056793
InChI Key WDQPAMHFFCXSNU-UHFFFAOYSA-N
Synonym N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine
PubChem CID 2794
ChEBI CHEBI:3749
IUPAC Name N,5-bis(4-chlorophenyl)-3-propan-2-yliminophenazin-2-amine
SMILES CC(C)N=C1C=C2C(=NC3=CC=CC=C3N2C4=CC=C(C=C4)Cl)C=C1NC5=CC=C(C=C5)Cl

Specifications

Melting Point 219°C
Color Red-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 473.40
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Clofazimine
RTECSNumber SG1578000
TSCA No