missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Clofazimine, 98%
Chemical Identifiers
| CAS | 2030-63-9 |
|---|---|
| Molecular Formula | C27H22Cl2N4 |
| Molecular Weight (g/mol) | 473.4 |
| InChI Key | WDQPAMHFFCXSNU-UHFFFAOYSA-N |
| PubChem CID | 2794 |
| ChEBI | CHEBI:3749 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461760010
|
Thermo Scientific™
461760010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC461760050
|
Thermo Scientific™
461760050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2030-63-9 | |
| Glass Bottle | |
| 1g | |
| WDQPAMHFFCXSNU-UHFFFAOYSA-N | |
| 2794 | |
| 473.4 | |
| Clofazimine |
| 97.5% min. (HPLC) | |
| C27H22Cl2N4 | |
| Solubility in water: insoluble. Other solubilities: soluble in toluene,slightly soluble in ether,acetone,ethyl acetate | |
| 473.4 | |
| CHEBI:3749 | |
| 98% |
Safety and Handling
EINECSNumber : 217-980-2
RUO â Research Use Only