Learn More
New England Biolab Ltd CLIP-Surface™ 547 50 nmol DFS Item
Supplier:  New England Biolab Ltd S9233S
CLIP-Surface 547 is a photostable red fluorescent substrate that can be used to label CLIP-tag fusion proteins on the surface of living cells or in vitro.
Catalog No. 508112087
Only null left
Explore available promotions