missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Clinical Chemistry Analyzer Sample Cups
Specifications
Specifications
| Closure Type | Snap |
| Material | Polyethylene |
| Product Type | Sample Cup |
| Color | Red |
| For Use With (Equipment) | Roche cobas Mira™ analyzer |
| For Use With (Application) | Designed for use on automated instrumentation |