missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Clinical Chemistry Analyzer Sample Cups

Catalog No. 22170198
Change view
Click to view available options
Color:
Blue
Clear
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22170198 Yellow
22170197 Red
22170195 Blue
22170196 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22170198 Supplier Globe Scientific Supplier No. 6500Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For various laboratory analyzers

Specifications

Color Yellow
Closure Type Snap
Material Polyethylene
Product Type Sample Cup
For Use With (Application) Designed for use on automated instrumentation
For Use With (Equipment) Roche cobas Mira™ analyzer