missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Clinical Chemistry Analyzer Sample Cups

Catalog No. 22170197
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Clear
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22170197 Red
22170198 Yellow
22170195 Blue
22170196 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. 22170197 missing translation for 'mfr' Globe Scientific missing translation for 'supplierNo' 6500R

For various laboratory analyzers

Specifications

Closure Type Snap
Material Polyethylene
Product Type Sample Cup
Color Red
For Use With (Equipment) Roche cobas Mira™ analyzer
For Use With (Application) Designed for use on automated instrumentation