missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin phosphate

Catalog No. AAJ63387MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63387MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63387MC Supplier Thermo Scientific Chemicals Supplier No. J63387MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Antibiotic U-28508E, effective against Gram-positive bacteria | CAS: 24729-96-2 | C18H34ClN2O8PS | 504.96 g/mol

Antibiotic U-28508E, effective against Gram-positive bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Type Clindamycin Phosphate
Physical Form Solid
CAS 24729-96-2
Melting Point 114°C
Color White
Quantity 100 mg
Molecular Formula C18H34ClN2O8PS
MDL Number MFCD07793328
Merck Index 14,2356
Solubility Information Freely soluble in water
InChI Key UFUVLHLTWXBHGZ-AVENPWRCSA-N
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl
IUPAC Name [(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate
Molecular Weight (g/mol) 504.96
PubChem CID 71314904
Formula Weight 504.96
Chemical Name or Material Clindamycin phosphate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 246-433-0
RTECSNumber GF2625000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only