missing translation for 'onlineSavingsMsg'
Learn More

Phosphate de clindamycine 97,0+%, TCI America™

Numéro de catalogue. p-7125367
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22575G 5 g
C225725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22575G Supplier TCI America Supplier No. C22575G

Chemical Identifiers

CAS 24729-96-2
Molecular Formula C18H34ClN2O8PS
Molecular Weight (g/mol) 504.96
MDL Number MFCD07793328
InChI Key UFUVLHLTWXBHGZ-AVENPWRCSA-N
Synonym Clindamycin 2-Dihydrogen Phosphate
PubChem CID 71314904
IUPAC Name [(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-méthyl-4-propylpyrrolidine-2-carbonyl]amino]propyle]-4,5-dihydroxy-2-méthylsulfanyloxane-3-yl] dihydrogène phosphate
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl

Specifications

Type Clindamycin Phosphate
Physical Form Poudre cristalline
Melting Point 114°C
Color Blanc/Jaune
Quantity 5 g
Formula Weight 504.96
Percent Purity ≥97.0% (T)
Chemical Name or Material Clindamycin Phosphate
RTECSNumber GF2625000
TSCA No