missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin Phosphate 97.0+%, TCI America™
Specifications
Specifications
| Type | Clindamycin Phosphate |
| Physical Form | Crystalline Powder |
| Melting Point | 114°C |
| Color | White/Yellow |
| Quantity | 5 g |
| Formula Weight | 504.96 |
| Percent Purity | ≥97.0% (T) |
| Grade | Not Available |
| Chemical Name or Material | Clindamycin Phosphate |
Safety and Handling
| RTECSNumber | GF2625000 |
| TSCA | No |