missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin Hydrochloride USP, Medisca™

$64.42 - $2,987.10
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048059
![]() |
N/A | 100 g |
Each for $487.64
|
|
|||||
18048060
![]() |
N/A | 500 g |
Each for $1,962.78
|
|
|||||
18048078
![]() |
N/A | 5 g |
Each for $64.42
|
|
|||||
18048079
![]() |
N/A | 25 g |
Each for $171.73
|
|
|||||
18048080
![]() |
N/A | 1 kg |
Each for $2,987.10
|
|
|||||
|
18048104
|
N/A | 10 g |
Each for $110.42
|
|
|||||
Description
Clindamycin Hydrochloride USP is a white or practically white, crystalline powder. It is odorless to having a faint mercaptan-like odor. Stable in the presence of air and light.
Chemical Identifiers
| 21462-39-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 21462-39-5 | |
| ≥ 800 μg |
| Crystalline Powder | |
| White | |
| USP |