Learn More
MilliporeSigma™ Supelco™ Click-On Inline Super Clean™ Gas Purifiers
Simple installation and replacement of the Click-On Purifier eliminates the risk of damage to gas line connections caused by overtightening
Supplier: MilliporeSigma™ Supelco™ 28862U
Description
- Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors
- Click-on Connectors are not included and must be purchased before initial use
Chemical Identifiers
| 194 mm | |
| Stainless Steel | |
| 1 Ea. | |
| 32 mm |
Specifications
| 194 mm | |
| 1 Ea. | |
| 32 mm |
| Stainless Steel | |
| Oxygen Trap |