missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ ClearVue™ Mountants
Specifications
Specifications
| Color | Light yellow |
| Product Type | Mountant |
| Quantity | 1 pt. |
| Solution Type | Mountant, Toluene-based |
| Formulation | Toluene, Acrylic Resin |
| Form | Liquid |