missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Clearify Clearing Agent

Catalog No. STLCACLEGAL
Change view
Click to view available options
Product Type:
Clearing Agent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
STLCACLEGAL Clearing Agent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLCACLEGAL Supplier StatLab™ Supplier No. CACLEGAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or inhaled.

Clearify is an ideal clearing agent, coverslipping medium, deparaffinizer, and xylene alternative.

  • Readily biodegradable and low-hazard
  • Non-carcinogenic, non-dermatologic, and non-corrosive
  • Looks and smells like water, but works like xylene

Specifications

Color Clear
Product Type Clearing Agent
Quantity 1 gal./Bottle, 4 Bottles/Cs.
For Use With (Application) Tissue Clearing
Form Liquid