missing translation for 'onlineSavingsMsg'
Learn More

Clear Lifting Tape

Catalog No. S69017
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S69017 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S69017 Supplier Customer Channels Group (Ccg) Supplier No. 5564

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crystal-clear tape for lifting fiber evidence.

  • Uniform adhesion
  • Proper clarity
  • No streaks or air bubbles
  • W × L: 2 in. × 30 ft. (5cm × 9.1m)

Specifications

Color Clear
For Use With (Application) For lifting fiber evidence
Product Type Lifting Tape