missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Clear Gate Valve Lockout
Description
Gate valve lockout devices deny access and cover the valve handle. This device's unique knockout center fits OSY or rising stem valves.
- Device is made of extremely rugged injection-molded polypropylene for superior impact and chemical resistance.
- Supplied with English, Spanish and French danger labels.
- Cover the valve handle with the gate valve lockout device and apply a personal padlock and lockout tag to ensure employee safety.
- Accepts padlock shackles up to 3/8 in. in diameter.
- Transparent gate valve covers allow visual inspection of the valve position and condition without having to remove the lockout device.
Specifications
Specifications
| Product Type | Lockout Device |
| Color | Clear |
| Material | Polycarbonate |
| Certifications/Compliance | OSHA 1910.147 |
| Description | Gate Valve Lockout |
| For Use With (Equipment) | Valve Handle: 2.5 to 5 in. |
| Includes | Attachable Cleat for Circuit Breaker Lockout Device |
| Quantity | 1 Ea. |