missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Cleanroom Products Clear Cleanroom Trash Bag
A convenient way to hold cleanroom generated waste.
$642.63 - $1076.41
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Certifications/Compliance | CoC |
| Closure Type | None |
| Color | Clear |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
|
17-010-249
|
Keystone Cleanroom Products
3440PPB25 |
33 gal. |
Case of 250 Each for $1,076.41
|
|
|||||
|
17-010-251
|
Keystone Cleanroom Products
3660CPPB25 |
55 gal. |
Case of 100 Each for $642.63
|
|
|||||
Description
- Cleanroom packed into sealed poly bag packages to prevent the introduction of foreign particulate contamination into the controlled production environments
- Each poly bag package is labeled with a Lot number for complete traceability
- 25 per sealed poly bag
- Eliminate the introduction of cardboard and cellulose packaging into the cleanroom
- Simply wipe the outer packaging instead of each bag prior to the introduction into the cleanroom
Specifications
| Not Autoclavable | |
| None | |
| Single-use | |
| LDPE | |
| 2 mil |
| CoC | |
| Clear | |
| Cleanroom Waste | |
| Non-sterile | |
| Cleanroom Bags |