Learn More
Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
Perform quantitative and qualitative solid phase immunoassays with these nonsterile 96-well plates, available in a variety of binding surfaces.
Specifications
| Barcode | No Barcode |
|---|---|
| Format | Standard |
| Lid | Without Lid |
| Detection Method | Colorimetric |
| Array | 8 x 12 |
Alert:
Each lot is functionally tested for IgG binding.
| Catalog Number | Mfr. No. | Surface Treatment | Certified | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Surface Treatment | Certified | Price | Quantity & Availability | ||||
|
12565261
|
Thermo Scientific™
430341 |
MaxiSorp | No |
N/A
|
|
||||
|
12565266
|
Thermo Scientific™
446140 |
PolySorp | No |
N/A
|
|
||||
|
12565267
|
Thermo Scientific™
446612 |
MaxiSorp | Yes |
N/A
|
|
||||
Description
Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of multiple surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific™ PolySorp™ for adsorption of hydrophobic molecules
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays
Ordering Information:
Please note that availability may vary by country. 446612 includes certificate.
Specifications
| No Barcode | |
| Without Lid | |
| 8 x 12 | |
| Clear | |
| Polystyrene | |
| 250 μL | |
| Round, C Bottom | |
| 5 | |
| C96 |
| Standard | |
| Colorimetric | |
| 60 Cases | |
| Microplate | |
| 96 | |
| Non-sterile | |
| 60 | |
| None |