missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One μClear™ Bottom 96-well Polystyrene Microplates

Description
- Non-Pyrogenic
- Material: Polystyrene
- Free of detectable DNase, RNase, human DNA
- Traced way back to production through defined LOT number system
- More suitable for non polar proteins and peptides
- Medium binding
Certifications
DIN ISO 9001
Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | Black |
| Product Type | Assay Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Show More |