Learn More
Alkali Scientific Western Blot Boxes, 6 X 4 X 1 1/4"", Min Volume 11mL, Clear, 5/pk
Catalog No. 502335982
Encompass
Western blot boxes and containers are available in clear. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90 and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. 502335982 Supplier Alkali Scientific Supplier No. BC120017
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Western blot boxes and containers are available in clear. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90 and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.