missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One μClear™ 96-well, Advanced TC Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Supplier: Greiner Bio-One 655986
Description
Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.
- Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
- For use with serum reduced and serum-free media
- Allows differentiation of semi-adherent cells
- Non-pyrogenic and non-cytotoxic
- Homogenous cell growth
- Higher transgene activity
- μClear coated well bottom
Specifications
| No | |
| Standard | |
| 34 mm2 | |
| Not Autoclavable | |
| Black, Clear Bottom | |
| 392 μL | |
| 25 to 340 μL | |
| Advanced TC | |
| 4800 x g (swinging bucket rotor) | |
| 8 | |
| Chimney |
| Free of detectable DNase/RNase, human DNA and pyrogens; Contents non-cytotoxic | |
| With Lid | |
| 8 x 12 | |
| 8/Pk., 32/Cs. | |
| Polystyrene | |
| 96 | |
| Sterile | |
| Flat Bottom (F-well) | |
| 32 | |
| 32/Cs. | |
| 128 x 85 x 14 mm |