missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ BioClean™ Chemotherapy Protective Apron with Sleeves BCAS

Catalog No. 19668250
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19668250 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Please see product validation pack or contact Ansell customer service for specific data on use of garments with cytotoxic drugs. Garments used for protection against such drugs must be selected specifically for the type of chemicals used.
Catalog No. 19668250 Supplier Ansell™ Supplier No. BCASL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please see product validation pack or contact Ansell customer service for specific data on use of garments with cytotoxic drugs. Garments used for protection against such drugs must be selected specifically for the type of chemicals used.

Ansell™ BioClean™ Chemotherapy Protective Apron with Sleeves BCAS provides protection against a range of cytotoxic drugs.

  • Tested against permeation standard
  • ASTM F739-12
  • Ultrasonically bonded seams with protective tape
  • Tie tapes at rear
  • Adjustable neck fastening
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
  • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
  • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
  • Tested against standard EN 14325 protective clothing against chemicals
  • Tested against permeation standard ASTM F739-12
  • Processed in an NEBB certified ISO Class 4 environment
  • Tested against standards: EN 530, Abrasion resistance; EN 6530, Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength

Specifications

Product Type Chemotherapy Protective Apron
For Use With (Application) Chemotherapy Protective
Size Large
Material Polyethylene, Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Seam Type Ultrasonically Bonded with Protective Tape
Sleeve Style Long Sleeve
Certifications/Compliance ISO 9001:2015, (EU)2016/425, EN ISO 13034:2005 + A1:2009, EN 14325, ASTM F739-12, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2

Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.