Learn More
Texwipe™ CleanStep™ Frames for Mat System
Provide an excellent surface for adhering mats
$274.89 - $344.32
| Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Price | Quantity | ||||
|
18-318-002
|
Texwipe™
FW1937 |
White | 19 x 37 in. |
Each for $274.89
|
|
||||
|
18-318-006
|
Texwipe™
FW3747 |
White | 37 x 47 in. |
Each for $344.32
|
|
||||
|
18-318-004
|
Texwipe™
FW2646 |
White | 26 x 46 in. |
Each for $329.10
|
|
||||
Description
- Made of polystyrene
- Lightweight, durable construction
- Nonslip design
- Cleaned with IPA
- Available in three sizes and black or white colors