Learn More
Purus™ Pure Comfort Cleanroom Socks
Purus™ Cleanroom Socks are designed for cleanroom use and comfort.
Supplier: Purus™ CRSLL
Description
- Available in both disposable and launderable styles
- Made from 100% polyester
- Cleanroom compatible
- Comfortable for extended use
- No particle generation
Specifications
| Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors | |
| Large | |
| Non-sterile | |
| Launderable Cleanroom Socks | |
| 200 Pair/Cs |
| Polyester | |
| Reusable | |
| 1 Pair/Bag, 200 Pair/Cs | |
| Class 10-1000 (ISO 4-6) |