missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURUS™ Cleanroom Shoes
Description
- Smooth surface for day-long comfort
- Antistatic
- Autoclavable
- Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanroom
- Non-slip sole
- Special conical design prevents spilled liquids from entering clog
- Heel strap
- Anatomic Heel Basin
- Toe Grabber
Specifications
Specifications
| Certifications/Compliance | ASTM F2913 |
| Color | White |
| For Use With (Application) | Pharmaceutical, Biotech, Semiconductor, Cleanrooms, Hospitals |
| Material | Thermoplastic Elastomer |
| Size | Large |
| Toe Style | Plain Toe |
| Packaging Quantity | 6/Cs. |
| Product Type | Cleanroom Shoes |