Learn More
Uniform Technology™ Cleanroom Boot Altessa Grid ISO 5 Cleanroom Boots
This cleanroom boot cover is ideal for contamination prevention in general, high-tech, semiconductor, optical manufacturing, and inspection applications.
Supplier:  Uniform Technology™ CBPX74WHXL
 View more versions of this product
Catalog No. 17779042
Only null left
Explore available promotions
Description
Description
  • Altessa Grid Fabric is made of 99% polyester to 1% carbon blend
  • Color-coded webbing across toe with strap
  • Webbing around ankle with side release buckle closure
  • 1 in. elastic and webbing top closure with snap adjustment
  • X-Stat ESD sole with twill reinforcement
  • Upper is sewn on with conductive thread
  • Meets standards required for use in ISO 5 (Class 100) to ISO 8 (Class 100,000) cleanrooms and other controlled environments
Specifications
Specifications
White
X-Large
Cleanroom Boots
Polyblend
Water Repellent
ISO 5 (Class 100)
Product Suggestions
Product Suggestions