Learn More
New Pig CleanMax™ Cleanroom Sterile Frock
Description
- A primary feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
- Suitable for use in the most critical sterile environments, such as those in pharmaceutical and life sciences industries.
- Made from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven.
- The material is designed to be low-linting and low-particulate, which is essential for maintaining the integrity of ISO Class 5 environments and meeting stringent industry standards.
- The fabric is engineered to be breathable, allowing for the passage of moisture vapor to help reduce heat stress and improve wearer comfort during long shifts.
- The sterile frocks are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
- The frock includes a front zipper with a storm flap and elastic wrists to ensure a secure fit, helping to prevent the ingress of contaminants and provide a high level of protection.
Specifications
Specifications
| Size | 4X-Large |