Learn More
New Pig CleanMax™ Cleanroom Sterile Frock
Catalog No. 17987895
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17987895 4X-Large
17987896 5X-Large
17987891 Large
17987890 Medium
17987889 Small
17987892 X-Large
17987893 2X-Large
17987894 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17987895 Supplier New Pig Supplier No. WPL30024XL
Only null left
Help protect yourself in cleanrooms from fluids, as well as certain pathogens and viruses, by wearing this sterile frock with bound seams
  • A primary feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
  • Suitable for use in the most critical sterile environments, such as those in pharmaceutical and life sciences industries.
  • Made from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven.
  • The material is designed to be low-linting and low-particulate, which is essential for maintaining the integrity of ISO Class 5 environments and meeting stringent industry standards.
  • The fabric is engineered to be breathable, allowing for the passage of moisture vapor to help reduce heat stress and improve wearer comfort during long shifts.
  • The sterile frocks are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
  • The frock includes a front zipper with a storm flap and elastic wrists to ensure a secure fit, helping to prevent the ingress of contaminants and provide a high level of protection.
Specifications
Size 4X-Large