Learn More
New Pig CleanMax™ Cleanroom Sterile Coverall
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile, bound-seam coverall with attached hood and boots
Supplier: New Pig WPL30044XL
Description
- A key feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
- Suitable for use in the most critical sterile environments, such as those in pharmaceutical and life sciences industries.
- Made from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven. This provides a strong barrier against particles, microorganisms, and liquid splashes.
- The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
- The fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
- The sterile coveralls are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
- The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles to provide a secure fit and prevent the ingress of contaminants.
Specifications
| 4X-Large |