Learn More
New Pig CleanMax™ Cleanroom Sterile Coverall
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile, bound-seam coverall with attached hood and boots
Supplier:  New Pig WPL3004XXXL
 View more versions of this product
Catalog No. 17-987-908
Only null left
Explore available promotions
Description
Description
  • A key feature is its sterile status, as it has been gamma-irradiated to a Sterility Assurance Level (SAL) of -6.
  • Suitable for use in the most critical sterile environments, such as those in pharmaceutical and life sciences industries.
  • Made from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven. This provides a strong barrier against particles, microorganisms, and liquid splashes.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • The fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • The sterile coveralls are typically folded in a specific manner to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
  • The coverall includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles to provide a secure fit and prevent the ingress of contaminants.
Specifications
Specifications
3X-Large
Product Suggestions
Product Suggestions