Learn More
New Pig CleanMax™ Cleanroom Sterile Coverall (25 Per Case)
Protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile coverall with bound seams and attached hood
Supplier:  New Pig WPL3008S
 View more versions of this product
Catalog No. 17-987-935
Only null left
Explore available promotions
Description
Description
  • Sterile and Gamma-Irradiated: This coverall is sterilized by gamma irradiation to a Sterility Assurance Level (SAL) of -6, making it suitable for the most critical sterile environments, such as those in pharmaceutical and life sciences.
  • Superior Particle and Liquid Barrier: The coverall provides an excellent barrier against particles, microorganisms, and non-hazardous liquid splashes.
  • Low-Linting and Low-Particulate: Designed to shed minimal lint and particles, it helps maintain the integrity of ISO Class 5-8 cleanroom environments.
  • Comfortable and Breathable: The fabric is engineered to be breathable, allowing for the passage of moisture vapor to help reduce heat stress for the wearer.
  • Secure and Aseptic Design: It features a front zipper with a storm flap, elastic wrists, and elastic ankles for a secure fit, and is aseptically folded for a clean gowning process.
  • Conveniently Packaged: Each case contains 25 sterile coveralls, making it a convenient and ready-to-use solution for cleanroom operations.
Specifications
Specifications
Small
Product Suggestions
Product Suggestions